Assay Details
Target Gene Details
Entrez Gene ID: | 319929 |
Gene Name: | RIKEN cDNA A630076J17 gene |
Gene Aliases: |
- |
Location: |
Chr.3:107230614-107234031 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A630076J17Rik | NM_001256174.1 | 3 | 312 | NP_001243103.1 |
| AA119196.1 | 3 | 289 | ||
| AI452133.1 | 1 | 355 |
Target Gene Details
Entrez Gene ID: | 246691 |
Gene Name: | prokineticin 1 |
Gene Aliases: |
EG-VEGF |
Location: |
Chr.3:107231916-107239707 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prok1 | AK044344.1 | 2 | 2187 | |
| BC042707.1 | 3 | 2183 |

Genomic Map