Assay Details
Target Gene Details
Entrez Gene ID: | 67157 |
Gene Name: | RIKEN cDNA 2610301B20 gene |
Gene Aliases: |
AI428449 |
Location: |
Chr.4:10874249-10899425 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610301B20Rik | NM_026005.3 | 6 | 1811 | NP_080281.3 |
| AI840411.1 | 1 | 323 | ||
| AK003887.1 | 1 | 1226 | ||
| AK011950.1 | 6 | 1740 | BAB27934.1 | |
| AK146358.1 | 6 | 1732 | BAE27110.1 | |
| BC058777.1 | 6 | 1701 | AAH58777.1 |

Genomic Map