Assay Details
Target Gene Details
Entrez Gene ID: | 73852 |
Gene Name: | DNA segment, Chr 3, ERATO Doi 751, expressed |
Gene Aliases: |
2810009O15Rik, 4930415G15Rik |
Location: |
Chr.3:41742606-41803292 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D3Ertd751e | NM_001099785.1 | 6 | 3295 | NP_001093255.1 |
| NM_001291048.1 | 5 | 888 | NP_001277977.1 | |
| NM_027271.1 | 5 | 3160 | NP_081547.1 | |
| NM_028667.3 | 7 | 3412 | NP_082943.2 | |
| AK012700.1 | 5 | 847 | BAB28420.1 | |
| AK138405.1 | 8 | 3517 | ||
| AK144064.1 | 6 | 3259 |

Genomic Map