Assay Details
Target Gene Details
Entrez Gene ID: | 212712 |
Gene Name: | special AT-rich sequence binding protein 2 |
Gene Aliases: |
mKIAA1034 |
Location: |
Chr.1:56793981-56978681 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Satb2 |

Genomic Map