Assay Details
Target Gene Details
Entrez Gene ID: | 13555 |
Gene Name: | E2F transcription factor 1 |
Gene Aliases: |
E2F-1, mKIAA4009 |
Location: |
Chr.2:154559400-154569892 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E2f1 | NM_001291105.1 | 7 | 1475 | NP_001278034.1 |
| NM_007891.5 | 7 | 1519 | NP_031917.1 | |
| AK017841.1 | 7 | 1467 | BAB30968.1 | |
| AK153968.1 | 7 | 1484 | BAE32285.1 | |
| AK220226.1 | 7 | 1216 | BAD90151.1 | |
| BC052160.1 | 7 | 1461 | AAH52160.2 | |
| L21973.1 | 7 | 1479 | AAA83217.1 |
More Information
Set Membership: |
|

Genomic Map