Assay Details
Target Gene Details
Entrez Gene ID: | 12421 |
Gene Name: | RB1-inducible coiled-coil 1 |
Gene Aliases: |
2900055E04Rik, 5930404L04Rik, Cc1, FIP200, LaXp180 |
Location: |
Chr.1:6214642-6276372 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rb1cc1 | NM_009826.4 | 15 | 3429 | NP_033956.2 |
| AB050017.2 | 15 | 3429 | BAB16846.2 | |
| AB070619.1 | 15 | 3429 | BAB85610.1 | |
| AB093216.1 | 12 | 2617 | BAC41400.1 | |
| AK041038.1 | 3 | 1217 | BAC30793.1 | |
| AK135130.1 | 7 | 1745 | ||
| BC150774.1 | 13 | 3029 |
More Information
Set Membership: |
|

Genomic Map