Assay Details
Target Gene Details
Entrez Gene ID: | 12042 |
Gene Name: | B cell leukemia/lymphoma 10 |
Gene Aliases: |
AI132454, BCL-10, C81403, CARMEN, CIPER, CLAP, ME10, cE10 |
Location: |
Chr.3:145924262-145934366 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl10 | NM_009740.2 | 3 | 1079 | NP_033870.1 |
| AF057701.1 | 3 | 963 | AAD15801.1 | |
| AF100339.1 | AAD16429.1 | |||
| AF134396.1 | 3 | 963 | AAD39148.1 | |
| AJ006289.1 | 3 | 960 | CAA06956.1 | |
| AK076082.1 | 3 | 991 | BAC36168.1 | |
| AK140179.1 | 3 | 951 | BAE24267.1 | |
| AK146236.1 | 3 | 985 | BAE27001.1 | |
| AK150883.1 | 3 | 980 | BAE29930.1 | |
| AK152563.1 | 3 | 980 | BAE31316.1 | |
| AK152847.1 | 3 | 976 | BAE31540.1 | |
| AK156890.1 | 3 | 1033 | BAE33885.1 | |
| AK168084.1 | 3 | 985 | BAE40057.1 | |
| AK169126.1 | 3 | 986 | BAE40905.1 | |
| AK172158.1 | 3 | 990 | BAE42852.1 | |
| BC024379.1 | 3 | 957 | AAH24379.1 |

Genomic Map