Assay Details
Target Gene Details
Entrez Gene ID: | 64436 |
Gene Name: | inositol polyphosphate-5-phosphatase E |
Gene Aliases: |
1200002L24Rik, 72kDa, mKIAA0123 |
Location: |
Chr.2:26396249-26409219 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inpp5e | NM_001290437.1 | 11 | 3326 | NP_001277366.1 |
| NM_033134.3 | 10 | 3657 | NP_149125.1 | |
| NR_110957.1 | 10 | 3744 | ||
| AF226683.1 | 10 | 3619 | AAF86957.1 | |
| AK154097.1 | 10 | 3658 | BAE32374.1 | |
| AK158135.1 | BAE34376.1 | |||
| AK170812.1 | ||||
| BC052717.1 | 10 | 3626 | AAH52717.1 | |
| BC080295.1 | 10 | 3622 | AAH80295.1 |
Target Gene Details
Entrez Gene ID: | 66865 |
Gene Name: | peptidase (mitochondrial processing) alpha |
Gene Aliases: |
1200002L24Rik, 4933435E07Rik, Alpha-MPP, P-55 |
Location: |
Chr.2:26389348-26397121 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pmpca | NM_173180.3 | 13 | 2762 | NP_775272.1 |
| AK004467.1 | 1 | 437 | ||
| AK004549.1 | 13 | 2762 | BAB23363.1 | |
| AK147110.1 | 13 | 2765 | BAE27682.1 | |
| AK158990.1 | 13 | 2730 | BAE34758.1 | |
| AK167975.1 | 13 | 2761 | ||
| AK169325.1 | 13 | 2757 | BAE41079.1 | |
| AK169342.1 | 13 | 2757 | BAE41094.1 |
More Information
Set Membership: |
|

Genomic Map