Assay Details
Target Gene Details
Entrez Gene ID: | 102954 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 10 |
Gene Aliases: |
AI853080, Dipp3a |
Location: |
Chr.X:6168696-6173044 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt10 | NM_001031664.1 | 2 | 769 | NP_001026834.1 |
| BC055771.1 | 2 | 726 | AAH55771.1 | |
| CX209897.1 |
More Information
Set Membership: |
|

Genomic Map