Assay Details
Target Gene Details
Entrez Gene ID: | 320940 |
Gene Name: | ATPase, class VI, type 11C |
Gene Aliases: |
A330005H02Rik, AI315324, Ig |
Location: |
Chr.X:60223283-60404859 on Build GRCm38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp11c | NM_001001798.2 | 29 | 5445 | NP_001001798.1 |
| NM_001037863.1 | 30 | 5550 | NP_001032952.1 | |
| BC106087.1 | 30 | 5529 | AAI06088.1 |
More Information
Set Membership: |
|

Genomic Map