Assay Details
Target Gene Details
Entrez Gene ID: | 56550 |
Gene Name: | ubiquitin-conjugating enzyme E2D 2A |
Gene Aliases: |
1500034D03Rik, Ubc2e, Ube2d2, ubc4 |
Location: |
Chr.18:35771559-35807172 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2d2a | NM_019912.2 | 7 | 2131 | NP_064296.1 |
| AK075844.1 | 1 | 223 | ||
| AK136197.1 | 7 | 2069 | BAE22869.1 | |
| AK139623.1 | 7 | 2097 | BAE24085.1 | |
| AK147568.1 | 6 | 7817 | ||
| AK162093.1 | 7 | 1936 | BAE36719.1 | |
| AK172088.1 | 7 | 1958 | ||
| BE688817.1 | 1 | 355 | ||
| U62483.1 | 7 | 2087 | AAB05772.1 |
More Information
Set Membership: |
|

Genomic Map