Assay Details
Target Gene Details
Entrez Gene ID: | 22154 |
Gene Name: | tubulin, beta 5 class I |
Gene Aliases: |
AA408537, AI596182, B130022C14Rik, M(beta)5 |
Location: |
Chr.17:35833920-35838301 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubb5 | NM_011655.5 | 4 | 2150 | NP_035785.1 |
| AK080778.1 | 1 | 294 | ||
| AK088137.1 | 1 | 316 | ||
| AK089071.1 | 1 | 338 | ||
| AK146567.1 | 4 | 2082 | BAE27265.1 | |
| AK169210.1 | 4 | 2081 | BAE40982.1 |
More Information
Set Membership: |
|

Genomic Map