Assay Details
Target Gene Details
Entrez Gene ID: | 271144 |
Gene Name: | ankyrin repeat and death domain containing 1B |
Gene Aliases: |
9330128J19Rik |
Location: |
Chr.13:96415030-96471354 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankdd1b | NM_001042714.1 | 14 | 1610 | NP_001036179.1 |
| AK033386.1 | 8 | 850 | ||
| AK033960.1 | 7 | 804 | BAC28528.1 | |
| AK084814.1 | 3 | 242 | ||
| BC112417.1 | 14 | 1607 |

Genomic Map