Assay Details
Target Gene Details
Entrez Gene ID: | 328563 |
Gene Name: | apolipoprotein L 11b |
Gene Aliases: |
A330102K04Rik |
Location: |
Chr.15:77633951-77643286 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apol11b | NM_001143686.1 | 4 | 1843 | NP_001137158.1 |
| AK039744.1 | 3 | 1721 | ||
| AY662646.1 | 5 | 2076 |
More Information
Set Membership: |
|

Genomic Map