Assay Details
Target Gene Details
Entrez Gene ID: | 69696 |
Gene Name: | RIKEN cDNA 2310057N15 gene |
Gene Aliases: |
Krtap13-1 |
Location: |
Chr.16:88773181-88774206 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2310057N15Rik | NM_027170.1 | 1 | 647 | NP_081446.1 |
| AK009968.1 | 1 | 647 | BAB26618.1 | |
| BC104340.1 | AAI04341.1 | |||
| BC104341.1 | AAI04342.1 |
More Information
Set Membership: |
|

Genomic Map