Assay Details
Target Gene Details
Entrez Gene ID: | 67847 |
Gene Name: | synuclein, alpha interacting protein (synphilin) |
Gene Aliases: |
2810407O15Rik, 4933427B05Rik, BB104594, SYPH1 |
Location: |
Chr.18:52767811-52915935 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sncaip | NM_001199151.1 | NP_001186080.1 | ||
| NM_001199153.1 | NP_001186082.1 | |||
| NM_001199154.1 | NP_001186083.1 | |||
| NM_026408.4 | NP_080684.3 | |||
| AF348447.1 | AAK30156.1 | |||
| AK016941.1 | BAB30511.1 | |||
| AK044974.1 | BAC32166.1 | |||
| AK081942.1 | ||||
| AK082079.1 | BAC38405.1 | |||
| AK132346.1 | BAE21118.1 | |||
| AK160111.1 | BAE35635.1 | |||
| BC098489.1 | AAH98489.1 | |||
| BC130225.1 |
More Information
Set Membership: |
|

Genomic Map