Assay Details
Target Gene Details
Entrez Gene ID: | 67011 |
Gene Name: | methyltransferase like 6 |
Gene Aliases: |
1600013P15Rik, AI467521, AU020711 |
Location: |
Chr.14:31460360-31495021 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mettl6 | NM_025907.3 | 6 | 1768 | NP_080183.1 |
| AK082949.1 | 6 | 1824 | ||
| AK133735.1 | ||||
| BC030449.1 | 6 | 1769 | AAH30449.1 |
More Information
Set Membership: |
|

Genomic Map