Assay Details
Target Gene Details
Entrez Gene ID: | 332131 |
Gene Name: | keratin 78 |
Gene Aliases: |
2310030B04Rik, Kb40 |
Location: |
Chr.15:101946001-101996609 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt78 | NM_212487.4 | 9 | 3289 | NP_997652.4 |
| AK009535.1 | 1 | 280 | ||
| AK146611.1 | 10 | 2593 |

Genomic Map