Assay Details
Target Gene Details
Entrez Gene ID: | 320191 |
Gene Name: | hook homolog 3 (Drosophila) |
Gene Aliases: |
5830454D03Rik, AI317159, E330005F07Rik |
Location: |
Chr.8:26021421-26119224 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hook3 | NM_207659.3 | 22 | 2382 | NP_997542.1 |
| AK042180.1 | 9 | 866 | BAC31191.1 | |
| AK084594.1 | 5 | 479 | BAC39222.1 | |
| AK137723.1 | 12 | 1118 | BAE23477.1 | |
| AK145149.1 | 22 | 2352 | BAE26261.1 | |
| AK170858.1 | 22 | 2327 | ||
| AY223806.1 | AAO43109.1 | |||
| BC090960.1 | 23 | 2298 | AAH90960.1 |
More Information
Set Membership: |
|

Genomic Map