Assay Details
Target Gene Details
Entrez Gene ID: | 434280 |
Gene Name: | predicted gene 5607 |
Gene Aliases: |
EG434280 |
Location: |
Chr.8:12385771-12436732 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm5607 | NR_027975.2 | |||
| BC066076.1 | AAH66076.1 |

Genomic Map