Assay Details
Target Gene Details
Entrez Gene ID: | 270160 |
Gene Name: | RAB39, member RAS oncogene family |
Gene Aliases: |
C230094F14Rik, Rab39a |
Location: |
Chr.9:53684110-53706232 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab39 | NM_175562.3 | NP_780771.1 | ||
| AB232635.1 | BAF02897.1 | |||
| AK049038.1 | BAC33522.1 | |||
| AK053487.1 | BAC35399.1 | |||
| BC049787.1 | AAH49787.1 |
More Information
Set Membership: |
|

Genomic Map