Assay Details
Target Gene Details
Entrez Gene ID: | 79202 |
Gene Name: | tumor necrosis factor receptor superfamily, member 22 |
Gene Aliases: |
2810028K06Rik, C130035G06Rik, SOBa, Tnfrh2, Tnfrsf1al2, mDcTrailr2 |
Location: |
Chr.7:143634808-143649752 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf22 | NM_023680.3 | 7 | 621 | NP_076169.2 |
| AJ278265.1 | 7 | 579 | CAC16406.1 | |
| AK012838.1 | 7 | 622 | BAB28502.2 | |
| AY046551.1 | 6 | 575 | AAL05073.1 | |
| AY165626.1 | 7 | 644 | AAN87806.1 | |
| AY165627.1 | 6 | 599 | AAN87807.1 |
More Information
Set Membership: |
|

Genomic Map