Assay Details
Target Gene Details
Entrez Gene ID: | 232087 |
Gene Name: | methionine adenosyltransferase II, alpha |
Gene Aliases: |
D630045P18Rik |
Location: |
Chr.6:72432796-72439608 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mat2a | NM_145569.4 | 9 | 2428 | NP_663544.1 |
| AK034757.1 | 9 | 2429 | BAC28823.1 | |
| AK052764.1 | 9 | 2427 | BAC35139.1 | |
| AK144189.1 | 9 | 2431 | BAE25756.1 | |
| AK145407.1 | 9 | 2427 | BAE26420.1 | |
| AK151281.1 | 8 | 3012 | BAE30267.1 | |
| AK160869.1 | 8 | 3012 | ||
| AK164137.1 | 9 | 2427 | BAE37643.1 | |
| AK166671.1 | 9 | 2427 | BAE38932.1 | |
| AK168155.1 | 9 | 2427 | BAE40120.1 | |
| AK169705.1 | 9 | 2427 | BAE41317.1 | |
| AK191201.1 | 1 | 76 | ||
| AK191379.1 | 1 | 92 | ||
| AK192240.1 | 1 | 76 | ||
| AK195580.1 | 1 | 97 | ||
| AK196623.1 | 1 | 87 | ||
| AK197103.1 | 1 | 76 | ||
| AK200816.1 | 1 | 85 | ||
| AK201922.1 | 1 | 87 | ||
| AK203805.1 | 1 | 76 | ||
| AK203913.1 | 1 | 94 | ||
| AK205713.1 | ||||
| AK209635.1 | ||||
| AK215987.1 | ||||
| BC058360.1 | 8 | 2989 | AAH58360.1 |
More Information
Set Membership: |
|

Genomic Map