Assay Details
Target Gene Details
Entrez Gene ID: | 16653 |
Gene Name: | v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog |
Gene Aliases: |
AI929937, K-ras, Ki-ras, Kras-2, Kras2, p21B, ras |
Location: |
Chr.6:145216699-145250300 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kras | NM_021284.6 | 5 | 1240 | NP_067259.4 |
| AK155064.1 | 5 | 1241 | BAE33023.1 | |
| AK164065.1 | 5 | 1241 | BAE37609.1 | |
| BC004642.1 | 5 | 1228 | AAH04642.1 | |
| BC010202.1 | 5 | 1228 | AAH10202.1 | |
| BC141051.1 | 4 | 1066 | ||
| U76425.1 | 1 | 479 | ||
| U76426.1 | 1 | 479 |
More Information
Set Membership: |
|

Genomic Map