Assay Details
Target Gene Details
Entrez Gene ID: | 67059 |
Gene Name: | Obg-like ATPase 1 |
Gene Aliases: |
2510025G09Rik, 2810405J23Rik, 2810409H07Rik, Gtpbp9, PTD004 |
Location: |
Chr.2:73092801-73214447 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ola1 | NM_025942.2 | NP_080218.1 | ||
| NM_030091.1 | NP_084367.1 | |||
| AK010817.1 | BAB27201.1 | |||
| AK012998.1 | ||||
| AK013055.1 | BAB28624.1 | |||
| AK017803.1 | ||||
| AK019142.1 | BAB31566.1 | |||
| AK159873.1 | BAE35446.1 | |||
| AK167458.1 | BAE39543.1 | |||
| AK170982.1 | ||||
| AK199282.1 | ||||
| AK205353.1 | ||||
| BC011318.1 | AAH11318.1 | |||
| BC029207.1 | AAH29207.1 | |||
| BG069761.2 |
More Information
Set Membership: |
|

Genomic Map