Assay Details
Target Gene Details
Entrez Gene ID: | 108699 |
Gene Name: | chimerin 1 |
Gene Aliases: |
0610007I19Rik, 0710001E19Rik, 1700112L09Rik, 2900046J01Rik, AI413815, ARHGAP2 |
Location: |
Chr.2:73610660-73775367 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chn1 | NM_001113246.2 | NP_001106717.2 | ||
| NM_001166603.1 | NP_001160075.1 | |||
| AB264772.1 | ||||
| BC054770.1 | AAH54770.1 | |||
| CA749754.1 |

Genomic Map