Assay Details
Target Gene Details
Entrez Gene ID: | 381484 |
Gene Name: | predicted gene 5150 |
Gene Aliases: |
- |
Location: |
Chr.3:15946871-16006371 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm5150 | NM_001081687.1 | NP_001075156.1 | ||
| DQ055451.1 |
More Information
Set Membership: |
|

Genomic Map