Assay Details
Target Gene Details
Entrez Gene ID: | 20195 |
Gene Name: | S100 calcium binding protein A11 (calgizzarin) |
Gene Aliases: |
EMAPI, Emap1, S100a14, S100c, cal |
Location: |
Chr.3:93520496-93526288 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a11 | NM_016740.3 | NP_058020.1 | ||
| AK002490.1 | BAB22139.1 | |||
| AK003889.1 | BAB23059.1 | |||
| AK164352.1 | BAE37754.1 | |||
| BC021916.1 | AAH21916.1 | |||
| BC086903.1 | AAH86903.1 | |||
| U41341.1 | AAA85181.1 |
More Information
Set Membership: |
|

Genomic Map