Assay Details
Target Gene Details
Entrez Gene ID: | 100043424 |
Gene Name: | predicted gene 14005 |
Gene Aliases: |
OTTMUSG00000015351 |
Location: |
Chr.2:128298663-128429351 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm14005 | NR_028589.1 | |||
| NR_028590.1 | ||||
| NR_028591.1 | ||||
| AK144927.1 | ||||
| AK157218.1 | BAE34005.1 |

Genomic Map