Assay Details
Target Gene Details
Entrez Gene ID: | 241636 |
Gene Name: | transglutaminase 6 |
Gene Aliases: |
TGM3L, TGY |
Location: |
Chr.2:130112416-130154232 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgm6 | NM_001289747.1 | 11 | 1706 | NP_001276676.1 |
| NM_001289748.1 | 10 | 1543 | NP_001276677.1 | |
| NM_001289749.1 | 8 | 1200 | NP_001276678.1 | |
| NM_177726.4 | 10 | 1562 | NP_808394.1 | |
| AK037229.1 | 10 | 1552 | BAC29768.1 | |
| AY159126.1 | 10 | 1552 | AAO17786.1 | |
| AY177606.1 | 8 | 1190 | AAO26186.1 | |
| AY177607.1 | 10 | 1543 | ||
| BC112420.1 | 10 | 1539 | ||
| BC113796.1 | 11 | 1683 |
More Information
Set Membership: |
|

Genomic Map