Assay Details
Target Gene Details
Entrez Gene ID: | 241638 |
Gene Name: | leucine zipper, putative tumor suppressor family member 3 |
Gene Aliases: |
Prosapip1 |
Location: |
Chr.2:130632765-130642844 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lzts3 | NM_001291027.1 | NP_001277956.1 | ||
| NM_001291028.1 | NP_001277957.1 | |||
| NM_197945.4 | NP_922936.3 | |||
| AK148251.1 | ||||
| AK161711.1 | BAE36545.1 | |||
| BC058280.1 | AAH58280.1 | |||
| BY258870.1 | ||||
| DN175861.1 |

Genomic Map