Assay Details
Target Gene Details
Entrez Gene ID: | 319929 |
Gene Name: | RIKEN cDNA A630076J17 gene |
Gene Aliases: |
- |
Location: |
Chr.3:107230614-107234031 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A630076J17Rik | NM_001256174.1 | NP_001243103.1 | ||
| AA119196.1 |

Genomic Map