Assay Details
Target Gene Details
Entrez Gene ID: | 228731 |
Gene Name: | NK2 homeobox 4 |
Gene Aliases: |
1700001P03Rik, Nkx-2.4, tinman |
Location: |
Chr.2:147083337-147085439 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkx2-4 | NM_023504.1 | 1 | 30 | NP_075993.1 |
More Information
Set Membership: |
|

Genomic Map