Assay Details
Target Gene Details
Entrez Gene ID: | 18472 |
Gene Name: | platelet-activating factor acetylhydrolase, isoform 1b, subunit 1 |
Gene Aliases: |
LIS-1, Lis1, MMS10-U, Mdsh, Ms10u, Pafaha |
Location: |
Chr.11:74673949-74724384 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pafah1b1 | NM_013625.4 | NP_038653.1 | ||
| NR_037610.2 | ||||
| AK144908.1 | BAE26129.1 | |||
| AK147207.1 | BAE27764.1 | |||
| AK147229.1 | ||||
| AK147237.1 | BAE27787.1 | |||
| AK147309.1 | BAE27837.1 | |||
| AK147507.1 | BAE27961.1 | |||
| AK147515.1 | BAE27965.1 | |||
| AK147533.1 | BAE27977.1 | |||
| AK147793.1 | BAE28141.1 | |||
| AK164008.1 | BAE37586.1 | |||
| AK167448.1 | BAE39534.1 | |||
| AK167824.1 | BAE39846.1 | |||
| AK168929.1 | BAE40742.1 | |||
| AK168960.1 | BAE40766.1 | |||
| AY189217.1 | AAO41716.1 | |||
| AY189218.1 | AAO41717.1 | |||
| BC014831.1 | AAH14831.1 | |||
| BC026141.1 | AAH26141.1 | |||
| L25108.1 | ||||
| L25109.1 | AAD23059.1 | |||
| U95116.1 | AAC04610.1 |

Genomic Map