Assay Details
Target Gene Details
Entrez Gene ID: | 18126 |
Gene Name: | nitric oxide synthase 2, inducible |
Gene Aliases: |
NOS-II, Nos-2, Nos2a, i-NOS, iNOS |
Location: |
Chr.11:78920859-78960232 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nos2 | NM_010927.3 | 15 | 1927 | NP_035057.1 |
| AF065919.1 | 15 | 1924 | AAC17914.1 | |
| AF065920.1 | 15 | 1924 | AAC17915.1 | |
| AF065921.2 | 15 | 1924 | AAC17916.2 | |
| AF065922.2 | 14 | 1797 | AAC17917.2 | |
| AF065923.2 | 14 | 1797 | AAC17918.2 | |
| AY090567.1 | 14 | 1747 | AAM11887.1 | |
| BC062378.1 | 15 | 1936 | AAH62378.1 | |
| M84373.1 | 15 | 1928 | AAA39834.1 | |
| M87039.1 | 15 | 1999 | AAA39315.1 | |
| M92649.1 | 15 | 1935 | ||
| U43428.1 | 15 | 1815 | AAC52356.1 |
More Information
Set Membership: |
|

Genomic Map