Assay Details
Target Gene Details
Entrez Gene ID: | 56016 |
Gene Name: | heme binding protein 2 |
Gene Aliases: |
SOUL |
Location: |
Chr.10:18540123-18546076 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hebp2 | NM_019487.3 | 1 | 271 | NP_062360.1 |
| AF117614.1 | 1 | 203 | AAD32097.1 | |
| AK038452.1 | 1 | 260 | ||
| AK078130.1 | 1 | 271 | BAC37141.1 | |
| BC006898.1 | 1 | 82 | AAH06898.1 |

Genomic Map