Assay Details
Target Gene Details
Entrez Gene ID: | 12228 |
Gene Name: | B cell translocation gene 3 |
Gene Aliases: |
ANA, tob5 |
Location: |
Chr.16:78359860-78377181 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Btg3 | NM_001297747.1 | NP_001284676.1 | ||
| NM_009770.3 | NP_033900.1 | |||
| AK144415.1 | BAE25879.1 | |||
| BC012705.1 | AAH12705.1 | |||
| BC094027.1 | AAH94027.1 | |||
| BC147656.1 | ||||
| BC147662.1 | ||||
| CK623612.1 | ||||
| D83745.1 | BAA29058.1 | |||
| Z72000.1 | CAA96519.1 |

Genomic Map