Assay Details
Target Gene Details
Entrez Gene ID: | 55960 |
Gene Name: | estrogen receptor-binding fragment-associated gene 9 |
Gene Aliases: |
AI835379, Rcas1 |
Location: |
Chr.15:44619169-44641542 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ebag9 | NM_019480.4 | NP_062353.3 | ||
| AF076524.1 | AAF23857.1 | |||
| AK004366.1 | BAB23277.1 | |||
| AK159499.1 | BAE35132.1 | |||
| AK160149.1 | BAE35657.1 | |||
| AY009091.1 | AAG41054.1 | |||
| BC108397.1 | AAI08398.1 | |||
| BC119118.1 | ||||
| BC119120.1 | ||||
| BC144740.1 |

Genomic Map