Assay Details
Target Gene Details
Entrez Gene ID: | 23970 |
Gene Name: | protein kinase C and casein kinase substrate in neurons 2 |
Gene Aliases: |
AI197433 |
Location: |
Chr.15:83375607-83464606 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pacsin2 | NM_001159509.1 | 1 | 96 | NP_001152981.1 |
| BC023502.1 | 1 | 60 | AAH23502.1 | |
| CJ118256.1 | 1 | 99 |
More Information
Set Membership: |
|

Genomic Map