Assay Details
Target Gene Details
Entrez Gene ID: | 245282 |
Gene Name: | apolipoprotein L 10A |
Gene Aliases: |
9030421J09Rik |
Location: |
Chr.15:77477047-77491069 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apol10a | NM_177744.4 | NP_808412.1 | ||
| AK033521.1 | BAC28339.1 |
More Information
Set Membership: |
|

Genomic Map