Assay Details
Target Gene Details
Entrez Gene ID: | 170654 |
Gene Name: | keratin associated protein 19-4 |
Gene Aliases: |
Krtap16-4 |
Location: |
Chr.16:88884786-88885089 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap19-4 | NM_130873.1 | 1 | 238 | NP_570943.1 |
| AF345294.1 | 1 | 238 | AAK52892.1 | |
| BC116200.1 | 1 | 238 | ||
| BC116201.1 | 1 | 238 |

Genomic Map