Assay Details
Target Gene Details
Entrez Gene ID: | 330671 |
Gene Name: | beta-1,4-N-acetyl-galactosaminyl transferase 4 |
Gene Aliases: |
BC038881, Gm1095, beta4GalNAcT4 |
Location: |
Chr.7:141061109-141072119 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4galnt4 | NM_177897.3 | NP_808565.2 | ||
| AB114827.1 | BAD15101.1 |
More Information
Set Membership: |
|

Genomic Map