Assay Details
Target Gene Details
Entrez Gene ID: | 108012 |
Gene Name: | adaptor-related protein complex 1, sigma 2 subunit |
Gene Aliases: |
1500012A13Rik, AI853648, EST1 |
Location: |
Chr.X:163907377-163933666 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap1s2 | NM_001290378.1 | NP_001277307.1 | ||
| NM_001290379.1 | NP_001277308.1 | |||
| NM_026887.3 | NP_081163.2 | |||
| AK005223.1 | BAB23892.1 | |||
| AK043776.1 | BAC31652.1 | |||
| AK045558.1 | BAC32418.1 | |||
| AK047724.1 | BAC33140.1 | |||
| AK053941.1 | BAC35599.1 | |||
| AK167690.1 | BAE39737.1 | |||
| BC046964.1 | AAH46964.1 |
More Information
Set Membership: |
|

Genomic Map