Assay Details
Target Gene Details
Entrez Gene ID: | 331491 |
Gene Name: | fibronectin type III domain containing 3C2 |
Gene Aliases: |
5031408O05Rik, Gm775 |
Location: |
Chr.X:106235246-106255372 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fnd3c2 | NM_001033424.3 | NP_001028596.2 | ||
| AK030291.1 | ||||
| AK146031.1 | BAE26844.1 |
More Information
Set Membership: |
|

Genomic Map