Assay Details
Target Gene Details
Entrez Gene ID: | 70062 |
Gene Name: | cancer/testis antigen 2 |
Gene Aliases: |
1700030B21Rik |
Location: |
Chr.X:65047644-65049017 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctag2 | NM_027302.2 | 1 | 83 | NP_081578.1 |
| AK006526.1 | 1 | 84 | BAB24635.1 | |
| BC049681.1 | 1 | 77 | AAH49681.1 |

Genomic Map