Assay Details
Target Gene Details
Entrez Gene ID: | 70237 |
Gene Name: | basic helix-loop-helix domain containing, class B9 |
Gene Aliases: |
2700087I09Rik |
Location: |
Chr.X:135841639-135891081 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bhlhb9 | NM_001098222.1 | 1 | 145 | NP_001091692.1 |
| NM_198161.2 | 1 | 145 | NP_937804.1 | |
| AK143861.1 | 1 | 68 | BAE25572.1 | |
| AK157023.1 | 1 | 99 | BAE33934.1 | |
| BB863588.1 | 1 | 143 | ||
| BY272101.1 | 1 | 145 | ||
| CF729268.1 | 1 | 85 | ||
| CK625652.1 | 1 | 134 |
More Information
Set Membership: |
|

Genomic Map