Assay Details
Target Gene Details
Entrez Gene ID: | 107569 |
Gene Name: | 5'-nucleotidase, cytosolic III |
Gene Aliases: |
1600024P05Rik, 2610206B05Rik, 3110004A18Rik, AI842776, Nt5c3a, Nt5y, PN-1, PN-I, PSN1, UMPH, Umph-1, Umph1, cN-III, lupin, p36 |
Location: |
Chr.6:56882400-56923932 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nt5c3 | NM_026004.3 | NP_080280.3 | ||
| AK011525.1 | BAB27677.2 | |||
| AK011894.1 | BAB27901.2 | |||
| AK208033.1 | ||||
| BY080680.1 |
More Information
Set Membership: |
|

Genomic Map