Assay Details
Target Gene Details
Entrez Gene ID: | 79264 |
Gene Name: | KRIT1, ankyrin repeat containing |
Gene Aliases: |
2010007K12Rik, A630036P20Rik, AA432855, AI450393, AI643869, BB155247, BB235701, Ccm1 |
Location: |
Chr.5:3803165-3844515 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 17 - Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krit1 | NM_001170552.1 | NP_001164023.1 | ||
| NM_030675.3 | NP_109600.2 | |||
| NR_033173.1 | ||||
| AF306509.1 | AAG18456.1 | |||
| AF310134.1 | AAG47775.1 | |||
| AK041574.1 | BAC30991.1 | |||
| AK085107.1 | ||||
| AK144667.1 | ||||
| AY328895.1 | AAQ92980.1 | |||
| AY464945.1 | AAR24089.1 | |||
| BC054819.1 | AAH54819.1 | |||
| BC085252.1 | AAH85252.1 | |||
| BC141042.1 |
More Information
Set Membership: |
|

Genomic Map