Assay Details
Target Gene Details
Entrez Gene ID: | 381820 |
Gene Name: | small integral membrane protein 10 like 1 |
Gene Aliases: |
2700089E24Rik, AI662692 |
Location: |
Chr.6:133104914-133110899 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smim10l1 | NM_001163445.2 | 1 | 128 | NP_001156917.1 |
| NM_001271582.1 | 1 | 128 | NP_001258511.1 | |
| NM_001271583.1 | 1 | 128 | NP_001258512.1 | |
| NR_073368.1 | 1 | 128 | ||
| AK005205.1 | 1 | 70 | ||
| AK031684.1 | 1 | 107 | ||
| AK050460.1 | 1 | 80 | ||
| AK076076.1 | 1 | 102 | ||
| AK077261.1 | 1 | 107 | ||
| AK170805.1 | 1 | 80 | BAE42040.1 | |
| BB868819.1 | 1 | 54 | ||
| BB873199.1 | 1 | 54 | ||
| BY708088.1 | 1 | 112 |

Genomic Map