Assay Details
Target Gene Details
Entrez Gene ID: | 665934 |
Gene Name: | predicted gene 7854 |
Gene Aliases: |
EG665934, ENSMUSG00000054483 |
Location: |
Chr.5:43151686-43235354 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm7854 | NR_028417.1 | |||
| AK041797.1 | BAC31070.1 | |||
| AK138349.1 | ||||
| AV378858.2 |

Genomic Map